siRNA Details
| Gene Symbol: Cd81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013087.1 | NP_037219.1 | 5 | 629 | ||
| GenBank mRNA | BC060583.1 | - | 5 | 460 | 6918296 | |
| FQ229194.1 | - | 3 | 209 | |||
| FQ231991.1 | - | 5 | 615 | |||
| FQ232886.1 | - | 5 | 617 | |||
| U19894.1 | - | 5 | 629 | |||

Cd81
Genomic Map