siRNA Details
| Gene Symbol: Vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013090.2 | NP_037222.2 | 5 | 870 | ||
| GenBank mRNA | BC092206.1 | - | 5 | 768 | 7313343 | |
| M24104.1 | - | 5 | 832 | |||
| U74621.1 | - | 4 | 1183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vamp1| Gene Symbol: Vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013090.2 | NP_037222.2 | 5 | 870 | ||
| GenBank mRNA | BC092206.1 | - | 5 | 768 | 7313343 | |
| M24104.1 | - | 5 | 832 | |||
| U74621.1 | - | 4 | 1183 | |||