siRNA Details
| Gene Symbol: Tnfrsf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013091.1 | NP_037223.1 | 3 | 491 | ||
| XM_006237334.2 | XP_006237396.1 | 2 | 726 | |||
| GenBank mRNA | AF329976.1 | - | 3 | 490 | ||
| AF329977.1 | - | 3 | 490 | |||
| AF329978.1 | - | 3 | 490 | |||
| AF329979.1 | - | 3 | 490 | |||
| AF329980.1 | - | 3 | 490 | |||
| AF329981.1 | - | 3 | 490 | |||
| BC086413.1 | - | 3 | 511 | 7110094 | ||
| M63122.1 | - | 3 | 491 | |||

Tnfrsf1a
Genomic Map