siRNA Details
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013095.3 | NP_037227.1 | 5, 6 | 853 | ||
| XM_008766216.1 | XP_008764438.1 | 3, 4 | 792 | |||
| XM_008766217.1 | XP_008764439.1 | 3, 4 | 518 | |||
| GenBank mRNA | BC064437.1 | - | 853 | 5599910 | ||
| DQ409172.1 | - | 2, 3 | 109 | |||
| U66479.1 | - | 5, 6 | 662 | |||

Smad3
Genomic Map