siRNA Details
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013097.2 | NP_037229.1 | 4 | 576 | ||
| XM_006245829.2 | XP_006245891.1 | 5 | 1256 | |||
| GenBank mRNA | AF397150.1 | - | 4 | 576 | ||
| AF397151.1 | - | 5 | 447 | |||
| BC089764.1 | - | 4 | 576 | 7130586 | ||
| X56060.1 | - | 4 | 429 | |||

Dnase1
Genomic Map