siRNA Details
| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278475.1 | NP_001265404.1 | 3 | 1556 | ||
| XM_008772362.1 | XP_008770584.1 | 3 | 2208 | |||
| GenBank mRNA | D88751.1 | - | 2 | 1848 | ||
| D88752.1 | - | 1 | 2273 | |||
More Information
Set Membership: |
|

Ptger1
Genomic Map