siRNA Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013101.3 | NP_037233.3 | 8 | 1067 | ||
| GenBank mRNA | AF110461.2 | - | 8 | 759 | ||
| L27057.1 | - | 8 | 1067 | |||
| L27062.1 | - | 2 | 597 | |||
| L36467.1 | - | 957 | ||||
| M25348.1 | - | 1 | 27 | |||
| M26715.1 | - | 2 | 331 | |||
| M26716.1 | - | 2 | 235 | |||
| M26717.1 | - | 2 | 299 | |||
| M28411.1 | - | 1 | 27 | |||

Pde4a
Genomic Map