siRNA Details
| Gene Symbol: Adrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013108.2 | NP_037240.2 | 2, 3 | 1453 | ||
| GenBank mRNA | M74716.1 | - | 2, 3 | 1452 | ||
| S56481.1 | - | 2, 3 | 1244 | |||
| S73473.1 | - | 2, 3 | 1283 | |||
More Information
Set Membership: |
|

Adrb3
Genomic Map