siRNA Details
| Gene Symbol: Pparg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013124.3 | NP_037256.1 | 1, 2 | 165 | ||
| GenBank mRNA | AB019561.1 | - | 1, 2 | 79 | ||
| AF156666.1 | - | 1, 2 | 127 | |||
| Y12882.2 | - | 1, 2 | 128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pparg| Gene Symbol: Pparg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013124.3 | NP_037256.1 | 1, 2 | 165 | ||
| GenBank mRNA | AB019561.1 | - | 1, 2 | 79 | ||
| AF156666.1 | - | 1, 2 | 127 | |||
| Y12882.2 | - | 1, 2 | 128 | |||