siRNA Details
| Gene Symbol: Pparg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145366.1 | NP_001138838.1 | 6 | 840 | ||
| NM_001145367.1 | NP_001138839.1 | 6 | 1018 | |||
| NM_013124.3 | NP_037256.1 | 5 | 815 | |||
| XM_006237009.2 | XP_006237071.1 | 6 | 811 | |||
| GenBank mRNA | AB011365.1 | - | 4 | 648 | ||
| AB019561.1 | - | 5 | 729 | |||
| AF156665.1 | - | 6 | 840 | |||
| AF156666.1 | - | 5 | 777 | |||
| AF246457.1 | - | 6 | 768 | |||
| AF246458.1 | - | 6 | 808 | |||
| Y12882.2 | - | 5 | 778 | |||

Pparg
Genomic Map