siRNA Details
| Gene Symbol: Pparg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145366.1 | NP_001138838.1 | 6 | 851 | ||
| NM_001145367.1 | NP_001138839.1 | 6 | 1029 | |||
| NM_013124.3 | NP_037256.1 | 5 | 826 | |||
| XM_006237009.2 | XP_006237071.1 | 6 | 822 | |||
| GenBank mRNA | AB011365.1 | - | 4 | 659 | ||
| AB019561.1 | - | 5 | 740 | |||
| AF156665.1 | - | 6 | 851 | |||
| AF156666.1 | - | 5 | 788 | |||
| AF246457.1 | - | 6 | 779 | |||
| AF246458.1 | - | 6 | 819 | |||
| Y12882.2 | - | 5 | 789 | |||

Pparg
Genomic Map