siRNA Details
| Gene Symbol: Scn5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160162.1 | NP_001153634.1 | 27 | 6148 | ||
| NM_013125.2 | NP_037257.1 | 28 | 6307 | |||
| GenBank mRNA | M27902.1 | - | 28 | 6307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn5a| Gene Symbol: Scn5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160162.1 | NP_001153634.1 | 27 | 6148 | ||
| NM_013125.2 | NP_037257.1 | 28 | 6307 | |||
| GenBank mRNA | M27902.1 | - | 28 | 6307 | ||