siRNA Details
| Gene Symbol: Il15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772363.1 | XP_008770585.1 | 5 | 519 | ||
| GenBank mRNA | BC097340.1 | - | 6 | 1369 | ||
| U69272.1 | - | 1107 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il15| Gene Symbol: Il15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772363.1 | XP_008770585.1 | 5 | 519 | ||
| GenBank mRNA | BC097340.1 | - | 6 | 1369 | ||
| U69272.1 | - | 1107 | ||||