siRNA Details
| Gene Symbol: Nr3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013131.1 | NP_037263.1 | 2 | 766 | ||
| GenBank mRNA | M36074.1 | - | 2 | 766 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr3c2| Gene Symbol: Nr3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013131.1 | NP_037263.1 | 2 | 766 | ||
| GenBank mRNA | M36074.1 | - | 2 | 766 | ||