siRNA Details
| Gene Symbol: Hmgcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013134.2 | NP_037266.2 | 20 | 3615 | ||
| XM_006231826.2 | XP_006231888.1 | 20 | 4023 | |||
| GenBank mRNA | BC064654.1 | - | 20 | 3615 | 5600023 | |
| FQ222434.1 | - | 1 | 408 | |||

Hmgcr
Genomic Map