siRNA Details
| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013136.1 | NP_037268.1 | 13 | 2021 | ||
| GenBank mRNA | BC078887.1 | - | 12 | 1886 | 7111717 | |
| M35862.1 | - | 13 | 2021 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mak| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013136.1 | NP_037268.1 | 13 | 2021 | ||
| GenBank mRNA | BC078887.1 | - | 12 | 1886 | 7111717 | |
| M35862.1 | - | 13 | 2021 | |||