siRNA Details
| Gene Symbol: Itpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013138.1 | NP_037270.1 | 4, 5 | 501 | ||
| XM_008772710.1 | XP_008770932.1 | 5, 6 | 565 | |||
| GenBank mRNA | GQ233031.1 | - | 4, 5 | 367 | ||
| L06096.1 | - | 4, 5 | 501 | |||

Itpr3
Genomic Map