siRNA Details
| Gene Symbol: Itpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013138.1 | NP_037270.1 | 5 | 528 | ||
| XM_008772710.1 | XP_008770932.1 | 6 | 592 | |||
| GenBank mRNA | GQ233031.1 | - | 5 | 394 | ||
| L06096.1 | - | 5 | 528 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpr3| Gene Symbol: Itpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013138.1 | NP_037270.1 | 5 | 528 | ||
| XM_008772710.1 | XP_008770932.1 | 6 | 592 | |||
| GenBank mRNA | GQ233031.1 | - | 5 | 394 | ||
| L06096.1 | - | 5 | 528 | |||