siRNA Details
| Gene Symbol: Gnai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013145.1 | NP_037277.1 | 8 | 1327 | ||
| GenBank mRNA | DQ120469.1 | - | 8 | 1201 | ||
| DQ120470.1 | - | 8 | 1201 | |||
| M17527.1 | - | 8 | 1327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnai1| Gene Symbol: Gnai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013145.1 | NP_037277.1 | 8 | 1327 | ||
| GenBank mRNA | DQ120469.1 | - | 8 | 1201 | ||
| DQ120470.1 | - | 8 | 1201 | |||
| M17527.1 | - | 8 | 1327 | |||