siRNA Details
| Gene Symbol: Cald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013146.2 | NP_037278.1 | 3 | 733 | ||
| XM_006236260.2 | XP_006236322.1 | 3 | 2283 | |||
| XM_006236261.2 | XP_006236323.1 | 4 | 804 | |||
| XM_006236262.2 | XP_006236324.1 | 4 | 957 | |||
| XM_008762763.1 | XP_008760985.1 | 4 | 807 | |||

Cald1
Genomic Map