siRNA Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013147.1 | NP_037279.1 | 6 | 831 | ||
| XM_008762873.1 | XP_008761095.1 | 6 | 1747 | |||
| GenBank mRNA | AY112736.1 | - | 6 | 659 | ||
| AY112737.1 | - | 6 | 659 | |||
| AY112738.1 | - | 6 | 659 | |||
| AY112739.1 | - | 6 | 659 | |||
| AY112740.1 | - | 6 | 659 | |||
| X62894.1 | - | 6 | 831 | |||

Clcn1
Genomic Map