siRNA Details
| Gene Symbol: Has2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013153.1 | NP_037285.1 | 4 | 2560 | ||
| GenBank mRNA | AF008201.1 | - | 4 | 2560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Has2| Gene Symbol: Has2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013153.1 | NP_037285.1 | 4 | 2560 | ||
| GenBank mRNA | AF008201.1 | - | 4 | 2560 | ||