siRNA Details
| Gene Symbol: Ide | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013159.1 | NP_037291.1 | 1, 2 | 110 | ||
| XM_006231313.2 | XP_006231375.1 | 1, 2 | 881 | |||
| GenBank mRNA | X67269.1 | - | 1, 2 | 110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ide| Gene Symbol: Ide | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013159.1 | NP_037291.1 | 1, 2 | 110 | ||
| XM_006231313.2 | XP_006231375.1 | 1, 2 | 881 | |||
| GenBank mRNA | X67269.1 | - | 1, 2 | 110 | ||