siRNA Details
| Gene Symbol: Mxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013160.2 | NP_037292.2 | 4 | 301 | ||
| XM_006231604.1 | XP_006231666.1 | 4 | 860 | |||
| XM_006231605.1 | XP_006231667.1 | 4 | 272 | |||
| XM_006231606.1 | XP_006231668.1 | 4 | 431 | |||
| GenBank mRNA | AF003008.1 | - | 4 | 254 | ||
| BC081824.1 | - | 4 | 301 | 7133853 | ||

Mxi1
Genomic Map