siRNA Details
| Gene Symbol: Mxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013160.2 | NP_037292.2 | 6 | 874 | ||
| XM_006231604.1 | XP_006231666.1 | 6 | 1433 | |||
| XM_006231605.1 | XP_006231667.1 | 6 | 845 | |||
| XM_006231606.1 | XP_006231668.1 | 6 | 1004 | |||
| GenBank mRNA | AF003008.1 | - | 6 | 825 | ||
| BC081824.1 | - | 6 | 874 | 7133853 | ||
| U44728.1 | - | 1 | 226 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001008542, NM_010847, NM_001008543, AK144219, BC064453, and others.

Mxi1
Genomic Map