siRNA Details
Gene Symbol: Il2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013163.1 | NP_037295.1 | 4 | 562 | ||
GenBank mRNA | FQ228242.1 | - | 594 | |||
M55049.1 | - | 4 | 562 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013163.1 | NP_037295.1 | 4 | 562 | ||
GenBank mRNA | FQ228242.1 | - | 594 | |||
M55049.1 | - | 4 | 562 |