siRNA Details
| Gene Symbol: Cckbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013165.2 | NP_037297.1 | 3 | 536 | ||
| GenBank mRNA | M99418.1 | - | 3 | 627 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cckbr| Gene Symbol: Cckbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013165.2 | NP_037297.1 | 3 | 536 | ||
| GenBank mRNA | M99418.1 | - | 3 | 627 | ||
Set Membership: |
|