siRNA Details
Gene Symbol: Cd3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013169.1 | NP_037301.1 | 2 | 271 | ||
GenBank mRNA | X53430.1 | - | 2 | 271 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013169.1 | NP_037301.1 | 2 | 271 | ||
GenBank mRNA | X53430.1 | - | 2 | 271 |