siRNA Details
| Gene Symbol: Scn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013178.1 | NP_037310.1 | 3, 4 | 847 | ||
| XM_006247559.2 | XP_006247621.1 | 4, 5 | 1197 | |||
| GenBank mRNA | M26643.1 | - | 3, 4 | 847 | ||
| Y17153.1 | - | 2, 3 | 398 | |||

Scn4a
Genomic Map