siRNA Details
| Gene Symbol: Scn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013178.1 | NP_037310.1 | 25 | 5402 | ||
| XM_006247559.2 | XP_006247621.1 | 26 | 5752 | |||
| GenBank mRNA | M26643.1 | - | 25 | 5402 | ||
| Y17153.1 | - | 24 | 4953 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn4a| Gene Symbol: Scn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013178.1 | NP_037310.1 | 25 | 5402 | ||
| XM_006247559.2 | XP_006247621.1 | 26 | 5752 | |||
| GenBank mRNA | M26643.1 | - | 25 | 5402 | ||
| Y17153.1 | - | 24 | 4953 | |||