siRNA Details
| Gene Symbol: Hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013185.3 | NP_037317.2 | 5 | 589 | ||
| GenBank mRNA | BC078890.1 | - | 5 | 589 | 7121407 | |
| M83666.1 | - | 5 | 511 | |||
| S74141.1 | - | 5 | 511 | |||
| X62345.1 | - | 5 | 540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hck| Gene Symbol: Hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013185.3 | NP_037317.2 | 5 | 589 | ||
| GenBank mRNA | BC078890.1 | - | 5 | 589 | 7121407 | |
| M83666.1 | - | 5 | 511 | |||
| S74141.1 | - | 5 | 511 | |||
| X62345.1 | - | 5 | 540 | |||