siRNA Details
| Gene Symbol: S100b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013191.1 | NP_037323.1 | 3 | 1425 | ||
| GenBank mRNA | M54919.1 | - | 3 | 1425 | ||
| X01090.1 | - | 3 | 1425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

S100b| Gene Symbol: S100b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013191.1 | NP_037323.1 | 3 | 1425 | ||
| GenBank mRNA | M54919.1 | - | 3 | 1425 | ||
| X01090.1 | - | 3 | 1425 | |||