siRNA Details
| Gene Symbol: Kcnj6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013192.2 | NP_037324.1 | 2 | 312 | ||
| XM_008768567.1 | XP_008766789.1 | 1 | 98 | |||
| GenBank mRNA | AB073753.1 | - | 2 | 312 | ||
| AB073754.1 | - | 2 | 312 | |||
| AB073755.1 | - | 2 | 312 | |||
| AB073756.1 | - | 2 | 312 | |||
| AY095171.1 | - | 1 | 101 | |||
| U21087.1 | - | 2 | 312 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001025584, NM_001025590, NM_001025585, NM_010606, U51126, and others.

Kcnj6
Genomic Map