siRNA Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013196.1 | NP_037328.1 | 5 | 784 | ||
| XM_006242151.2 | XP_006242213.1 | 4 | 1740 | |||
| XM_006242152.2 | XP_006242214.1 | 6 | 1012 | |||
| XM_006242153.2 | XP_006242215.1 | 6 | 727 | |||
| XM_006242154.2 | XP_006242216.1 | 6 | 781 | |||
| XM_006242156.2 | XP_006242218.1 | 5 | 611 | |||
| XM_008765676.1 | XP_008763898.1 | 6 | 804 | |||
| GenBank mRNA | M88592.1 | - | 5 | 784 | ||

Ppara
Genomic Map