siRNA Details
| Gene Symbol: Gfer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013222.2 | NP_037354.2 | 3 | 550 | ||
| GenBank mRNA | D30735.1 | - | 3 | 550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfer| Gene Symbol: Gfer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013222.2 | NP_037354.2 | 3 | 550 | ||
| GenBank mRNA | D30735.1 | - | 3 | 550 | ||