siRNA Details
Gene Symbol: Cebpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017095.1 | NP_058791.1 | 1 | 164 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
