siRNA Details
| Gene Symbol: Crp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017096.3 | NP_058792.1 | 2 | 235 | ||
| GenBank mRNA | AY310153.1 | - | 2129 | |||
| AY318961.1 | - | 2129 | ||||
| AY321318.1 | - | 2126 | ||||
| AY321324.1 | - | 2129 | ||||
| AY325196.1 | - | 2129 | ||||
| AY325218.1 | - | 2129 | ||||
| AY325232.1 | - | 2129 | ||||
| BC091157.1 | - | 303 | 7368286 | |||
| M83176.1 | - | 2 | 235 | |||

Crp
Genomic Map