siRNA Details
| Gene Symbol: Ctsc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017097.1 | NP_058793.1 | 2 | 319 | ||
| XM_008759636.1 | XP_008757858.1 | 2 | 393 | |||
| XM_008759637.1 | XP_008757859.1 | 2 | 392 | |||
| GenBank mRNA | D90404.1 | - | 2 | 319 | ||
| FQ218311.1 | - | 379 | ||||

Ctsc
Genomic Map