siRNA Details
| Gene Symbol: Ctsc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017097.1 | NP_058793.1 | 3 | 405 | ||
| GenBank mRNA | D90404.1 | - | 3 | 405 | ||
| FQ218311.1 | - | 465 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsc| Gene Symbol: Ctsc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017097.1 | NP_058793.1 | 3 | 405 | ||
| GenBank mRNA | D90404.1 | - | 3 | 405 | ||
| FQ218311.1 | - | 465 | ||||