siRNA Details
| Gene Symbol: Kcnj8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017099.4 | NP_058795.3 | 2, 3 | 549 | ||
| GenBank mRNA | AB043636.1 | - | 1, 2 | 379 | ||
| AB043637.1 | - | 1, 2 | 379 | |||
| BC078863.1 | - | 532 | 7128284 | |||
| D42145.1 | - | 2, 3 | 552 | |||

Kcnj8
Genomic Map