siRNA Details
| Gene Symbol: Plk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017100.1 | NP_058796.1 | 2 | 522 | ||
| GenBank mRNA | BC083926.1 | - | 2 | 498 | 7135428 | |
| U10188.1 | - | 2 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plk1| Gene Symbol: Plk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017100.1 | NP_058796.1 | 2 | 522 | ||
| GenBank mRNA | BC083926.1 | - | 2 | 498 | 7135428 | |
| U10188.1 | - | 2 | 522 | |||