siRNA Details
| Gene Symbol: FBXO25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012173.3 | NP_036305.2 | 8 | 1098 | ||
| NM_183420.1 | NP_904356.1 | 9 | 1148 | |||
| NM_183421.1 | NP_904357.1 | 9 | 1148 | |||
| XM_011534748.2 | XP_011533050.1 | 9 | 921 | |||
| XM_011534749.1 | XP_011533051.1 | 8 | 858 | |||
| XM_017013313.1 | XP_016868802.1 | 9 | 972 | |||
| XM_017013314.1 | XP_016868803.1 | 8 | 1050 | |||
| GenBank mRNA | AB070270.1 | - | 8 | 882 | ||
| AF174605.1 | - | 5 | 624 | |||
| AF419858.1 | - | 9 | 1148 | |||
| AK302334.1 | - | 10 | 823 | |||
| BC020249.2 | - | 7 | 840 | 4652811 | ||
| BC050393.1 | - | 9 | 1001 | 5769518 | ||

FBXO25
Genomic Map