siRNA Details
| Gene Symbol: Clcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017106.1 | NP_058802.1 | 12 | 2502 | ||
| GenBank mRNA | D50497.1 | - | 12 | 2502 | ||
| Z56277.1 | - | 12 | 2454 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcn5| Gene Symbol: Clcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017106.1 | NP_058802.1 | 12 | 2502 | ||
| GenBank mRNA | D50497.1 | - | 12 | 2502 | ||
| Z56277.1 | - | 12 | 2454 | |||