siRNA Details
| Gene Symbol: Ogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017107.2 | NP_058803.2 | 21 | 2747 | ||
| XM_006257056.1 | XP_006257118.1 | 20 | 2882 | |||
| XM_006257057.2 | XP_006257119.1 | 21 | 2824 | |||
| XM_006257058.1 | XP_006257120.1 | 20 | 2852 | |||
| GenBank mRNA | CK355423.1 | - | 2 | 235 | ||

Ogt
Genomic Map