siRNA Details
| Gene Symbol: Kcnh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017108.1 | NP_058804.1 | 4, 5 | 727 | ||
| GenBank mRNA | AB022697.1 | - | 4, 5 | 727 | ||
| AF073892.1 | - | 2, 3 | 215 | |||
| AJ007627.1 | - | 4, 5 | 592 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh3| Gene Symbol: Kcnh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017108.1 | NP_058804.1 | 4, 5 | 727 | ||
| GenBank mRNA | AB022697.1 | - | 4, 5 | 727 | ||
| AF073892.1 | - | 2, 3 | 215 | |||
| AJ007627.1 | - | 4, 5 | 592 | |||