siRNA Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017116.2 | NP_058812.1 | 21 | 2718 | ||
| GenBank mRNA | BC065306.1 | - | 21 | 2718 | 5621252 | |
| FQ209553.1 | - | 2 | 269 | |||
| FQ225985.1 | - | 2 | 273 | |||
| L09120.1 | - | 21 | 2404 | |||

Capn2
Genomic Map