siRNA Details
| Gene Symbol: Gzmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017119.2 | NP_058815.1 | 3 | 256 | ||
| GenBank mRNA | L19694.1 | - | 3 | 256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gzmk| Gene Symbol: Gzmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017119.2 | NP_058815.1 | 3 | 256 | ||
| GenBank mRNA | L19694.1 | - | 3 | 256 | ||