siRNA Details
| Gene Symbol: Hpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017122.1 | NP_058818.1 | 3 | 1253 | ||
| GenBank mRNA | D12573.1 | - | 3 | 1253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpca| Gene Symbol: Hpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017122.1 | NP_058818.1 | 3 | 1253 | ||
| GenBank mRNA | D12573.1 | - | 3 | 1253 | ||