siRNA Details
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017123.1 | NP_058819.1 | 3 | 471 | ||
GenBank mRNA | X55183.1 | - | 3 | 471 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017123.1 | NP_058819.1 | 3 | 471 | ||
GenBank mRNA | X55183.1 | - | 3 | 471 |