siRNA Details
| Gene Symbol: Inhba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017128.2 | NP_058824.1 | 2 | 320 | ||
| XM_006254001.2 | XP_006254063.1 | 3 | 3979 | |||
| XM_008771712.1 | XP_008769934.1 | 2 | 3897 | |||
| XM_008771713.1 | XP_008769935.1 | 4 | 5687 | |||
| XM_008771714.1 | XP_008769936.1 | 4 | 2467 | |||
| XM_008771715.1 | XP_008769937.1 | 4 | 2369 | |||
| XM_008771716.1 | XP_008769938.1 | 4 | 747 | |||
| GenBank mRNA | M37482.1 | - | 2 | 312 | ||

Inhba
Genomic Map